Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 15 - 75354114 - 75354114 [-] on 75354114 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-631 | MIMAT0003300 |
Stem-loop Details
Human
