Assay Details
Gene Family ID: | MIPF0000039, mir-34 |
Mature miRNA Sequence: | CAAUCACUAACUCCACUGCCAU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-34b-3p | MIMAT0004676 |
Pan troglodytes | ptr-miR-34b | MIMAT0008113 |
Pongo pygmaeus | ppy-miR-34b | MIMAT0015741 |
Rhesus monkey | mml-miR-34b | MIMAT0006174 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
