Assay Details
Gene Family ID: | MIPF0000039, mir-34 |
Mature miRNA Sequence: | CAAUCACUAACUCCACUGCCAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-34b-3p | MIMAT0004676 |
| Pan troglodytes | ptr-miR-34b | MIMAT0008113 |
| Pongo pygmaeus | ppy-miR-34b | MIMAT0015741 |
| Rhesus monkey | mml-miR-34b | MIMAT0006174 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map