Assay Details
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUCUGAAAUCGGUUA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-29a-3p | MIMAT0000086 | |
Mouse | mmu-miR-29a-3p | MIMAT0000535 | |
Rat | rno-miR-29a-3p | MIMAT0000802 | |
Callithrix jacchus | cja-miR-29a | MIMAT0049306 | |
Dasypus novemcinctus | dno-miR-29a-3p | MIMAT0047550 | |
Daubentonia madagascariensis | dma-miR-29a | MIMAT0049287 | |
View More Rows | |||
Dog | cfa-miR-29a | MIMAT0006626 | |
Guinea pig | cpo-miR-29a-3p | MIMAT0046897 | |
Horse | eca-miR-29a | MIMAT0012940 | |
Microcebus murinus | mmr-miR-29a | MIMAT0049131 | |
Nomascus leucogenys | nle-miR-29a | MIMAT0049441 | |
Otolemur garnettii | oga-miR-29a | MIMAT0049630 | |
Papio hamadryas | pha-miR-29a | MIMAT0049536 | |
Pteropus alecto | pal-miR-29a-3p | MIMAT0039802 | |
Rabbit | ocu-miR-29a-3p | MIMAT0048144 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Guinea pig
Horse
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Pteropus alecto
Rabbit

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-29a (v17)
Mouse: mmu-miR-29a (v17)
Rat: rno-miR-29a (v18)
Mouse: mmu-miR-29a (v17)
Rat: rno-miR-29a (v18)
