Assay Details
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUCUGAAAUCGGUUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-29a-3p | MIMAT0000086 |
Mouse | mmu-miR-29a-3p | MIMAT0000535 |
Rat | rno-miR-29a-3p | MIMAT0000802 |
Callithrix jacchus | cja-miR-29a | MIMAT0049306 |
Dasypus novemcinctus | dno-miR-29a-3p | MIMAT0047550 |
Daubentonia madagascariensis | dma-miR-29a | MIMAT0049287 |
View More Rows | ||
Dog | cfa-miR-29a | MIMAT0006626 |
Guinea pig | cpo-miR-29a-3p | MIMAT0046897 |
Horse | eca-miR-29a | MIMAT0012940 |
Microcebus murinus | mmr-miR-29a | MIMAT0049131 |
Nomascus leucogenys | nle-miR-29a | MIMAT0049441 |
Otolemur garnettii | oga-miR-29a | MIMAT0049630 |
Papio hamadryas | pha-miR-29a | MIMAT0049536 |
Pteropus alecto | pal-miR-29a-3p | MIMAT0039802 |
Rabbit | ocu-miR-29a-3p | MIMAT0048144 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Guinea pig
Horse
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Pteropus alecto
Rabbit

More Information
Related Products
miRBase Alias:
Human: hsa-miR-29a (v17)
Mouse: mmu-miR-29a (v17)
Rat: rno-miR-29a (v18)
Mouse: mmu-miR-29a (v17)
Rat: rno-miR-29a (v18)
