Assay Details
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUCUGAAAUCGGUUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-29a-3p | MIMAT0000086 |
| Mouse | mmu-miR-29a-3p | MIMAT0000535 |
| Rat | rno-miR-29a-3p | MIMAT0000802 |
| Callithrix jacchus | cja-miR-29a | MIMAT0049306 |
| Dasypus novemcinctus | dno-miR-29a-3p | MIMAT0047550 |
| Daubentonia madagascariensis | dma-miR-29a | MIMAT0049287 |
| View More Rows | ||
| Dog | cfa-miR-29a | MIMAT0006626 |
| Guinea pig | cpo-miR-29a-3p | MIMAT0046897 |
| Horse | eca-miR-29a | MIMAT0012940 |
| Microcebus murinus | mmr-miR-29a | MIMAT0049131 |
| Nomascus leucogenys | nle-miR-29a | MIMAT0049441 |
| Otolemur garnettii | oga-miR-29a | MIMAT0049630 |
| Papio hamadryas | pha-miR-29a | MIMAT0049536 |
| Pteropus alecto | pal-miR-29a-3p | MIMAT0039802 |
| Rabbit | ocu-miR-29a-3p | MIMAT0048144 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Guinea pig
Horse
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Pteropus alecto
Rabbit

Genomic Map