Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | UCGAGGAGCUCACAGUCUAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-151a-5p | MIMAT0004697 | |
Mouse | mmu-miR-151-5p | MIMAT0004536 | |
Rat | rno-miR-151-5p | MIMAT0000613 | |
Bovine | bta-miR-151-5p | MIMAT0003523 | |
Cricetulus griseus | cgr-miR-151-5p | MIMAT0023782 | |
Daubentonia madagascariensis | dma-miR-151 | MIMAT0049293 | |
View More Rows | |||
Dog | cfa-miR-151 | MIMAT0006615 | |
Goat | chi-miR-151-5p | MIMAT0035981 | |
Horse | eca-miR-151-5p | MIMAT0013004 | |
Otolemur garnettii | oga-miR-28b | MIMAT0049659 | |
Papio hamadryas | pha-miR-151 | MIMAT0049561 | |
Pig | ssc-miR-151-5p | MIMAT0013882 | |
Pongo pygmaeus | ppy-miR-151a-5p | MIMAT0015775 | |
Pteropus alecto | pal-miR-151-5p | MIMAT0040079 | |
Rabbit | ocu-miR-151-5p | MIMAT0048265 | |
Rhesus monkey | mml-miR-151-5p | MIMAT0006212 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Daubentonia madagascariensis
Dog
Goat
Horse
Otolemur garnettii
Papio hamadryas
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool B v3.0Megaplex™ RT Primers, Rodent Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Rodent MicroRNA B Cards v3.0TaqMan® Human MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-151-5p (v17)
Rat: rno-miR-151 (v18)
Bos taurus: bta-miR-151* (v18)
Pongo pygmaeus: ppy-miR-151-5p (v18)
Rat: rno-miR-151 (v18)
Bos taurus: bta-miR-151* (v18)
Pongo pygmaeus: ppy-miR-151-5p (v18)
