Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | UCGAGGAGCUCACAGUCUAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-151a-5p | MIMAT0004697 |
| Mouse | mmu-miR-151-5p | MIMAT0004536 |
| Rat | rno-miR-151-5p | MIMAT0000613 |
| Bovine | bta-miR-151-5p | MIMAT0003523 |
| Cricetulus griseus | cgr-miR-151-5p | MIMAT0023782 |
| Daubentonia madagascariensis | dma-miR-151 | MIMAT0049293 |
| View More Rows | ||
| Dog | cfa-miR-151 | MIMAT0006615 |
| Goat | chi-miR-151-5p | MIMAT0035981 |
| Horse | eca-miR-151-5p | MIMAT0013004 |
| Otolemur garnettii | oga-miR-28b | MIMAT0049659 |
| Papio hamadryas | pha-miR-151 | MIMAT0049561 |
| Pig | ssc-miR-151-5p | MIMAT0013882 |
| Pongo pygmaeus | ppy-miR-151a-5p | MIMAT0015775 |
| Pteropus alecto | pal-miR-151-5p | MIMAT0040079 |
| Rabbit | ocu-miR-151-5p | MIMAT0048265 |
| Rhesus monkey | mml-miR-151-5p | MIMAT0006212 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Daubentonia madagascariensis
Dog
Goat
Horse
Otolemur garnettii
Papio hamadryas
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map