Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | UCGAGGAGCUCACAGUCUAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-151a-5p | MIMAT0004697 |
| Mouse | mmu-miR-151-5p | MIMAT0004536 |
| Rat | rno-miR-151-5p | MIMAT0000613 |
| Bovine | bta-miR-151-5p | MIMAT0003523 |
| Cricetulus griseus | cgr-miR-151-5p | MIMAT0023782 |
| Daubentonia madagascariensis | dma-miR-151 | MIMAT0049293 |
| View More Rows | ||
| Dog | cfa-miR-151 | MIMAT0006615 |
| Goat | chi-miR-151-5p | MIMAT0035981 |
| Horse | eca-miR-151-5p | MIMAT0013004 |
| Otolemur garnettii | oga-miR-28b | MIMAT0049659 |
| Papio hamadryas | pha-miR-151 | MIMAT0049561 |
| Pig | ssc-miR-151-5p | MIMAT0013882 |
| Pongo pygmaeus | ppy-miR-151a-5p | MIMAT0015775 |
| Pteropus alecto | pal-miR-151-5p | MIMAT0040079 |
| Rabbit | ocu-miR-151-5p | MIMAT0048265 |
| Rhesus monkey | mml-miR-151-5p | MIMAT0006212 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Daubentonia madagascariensis
Dog
Goat
Horse
Otolemur garnettii
Papio hamadryas
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-151-5p (v17)
Rat: rno-miR-151 (v18)
Bos taurus: bta-miR-151* (v18)
Pongo pygmaeus: ppy-miR-151-5p (v18)
Rat: rno-miR-151 (v18)
Bos taurus: bta-miR-151* (v18)
Pongo pygmaeus: ppy-miR-151-5p (v18)

Genomic Map