Assay Details
Gene Family ID: | MIPF0000649, mir-1296 |
Mature miRNA Sequence: | UUAGGGCCCUGGCUCCAUCUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-1296-5p | MIMAT0005794 | |
| Bovine | bta-miR-1296 | MIMAT0009964 | |
| Callithrix jacchus | cja-miR-1296 | MIMAT0039512 | |
| Dasypus novemcinctus | dno-miR-1296-5p | MIMAT0048039 | |
| Goat | chi-miR-1296 | MIMAT0035938 | |
| Gorilla gorilla | ggo-miR-1296 | MIMAT0024172 | |
| View More Rows | |||
| Guinea pig | cpo-miR-1296-5p | MIMAT0047415 | |
| Horse | eca-miR-1296 | MIMAT0012890 | |
| Nomascus leucogenys | nle-miR-1296 | MIMAT0049452 | |
| Otolemur garnettii | oga-miR-1296 | MIMAT0049613 | |
| Pan troglodytes | ptr-miR-1296 | MIMAT0008015 | |
| Pig | ssc-miR-1296-5p | MIMAT0022964 | |
| Pongo pygmaeus | ppy-miR-1296 | MIMAT0016217 | |
| Rabbit | ocu-miR-1296-5p | MIMAT0048593 | |
Stem-loop Details
Human
Bovine
Callithrix jacchus
Dasypus novemcinctus
Goat
Gorilla gorilla
Guinea pig
Horse
Nomascus leucogenys
Otolemur garnettii
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit

Genomic Map