Assay Details
Gene Family ID: | MIPF0000649, mir-1296 |
Mature miRNA Sequence: | UUAGGGCCCUGGCUCCAUCUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1296-5p | MIMAT0005794 |
Bovine | bta-miR-1296 | MIMAT0009964 |
Callithrix jacchus | cja-miR-1296 | MIMAT0039512 |
Dasypus novemcinctus | dno-miR-1296-5p | MIMAT0048039 |
Goat | chi-miR-1296 | MIMAT0035938 |
Gorilla gorilla | ggo-miR-1296 | MIMAT0024172 |
View More Rows | ||
Guinea pig | cpo-miR-1296-5p | MIMAT0047415 |
Horse | eca-miR-1296 | MIMAT0012890 |
Nomascus leucogenys | nle-miR-1296 | MIMAT0049452 |
Otolemur garnettii | oga-miR-1296 | MIMAT0049613 |
Pan troglodytes | ptr-miR-1296 | MIMAT0008015 |
Pig | ssc-miR-1296-5p | MIMAT0022964 |
Pongo pygmaeus | ppy-miR-1296 | MIMAT0016217 |
Rabbit | ocu-miR-1296-5p | MIMAT0048593 |
Stem-loop Details
Human
Bovine
Callithrix jacchus
Dasypus novemcinctus
Goat
Gorilla gorilla
Guinea pig
Horse
Nomascus leucogenys
Otolemur garnettii
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
