Assay Details
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | ACCAUCGACCGUUGAGUGGACC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-181c-3p | MIMAT0017068 | |
| Rat | rno-miR-181c-3p | MIMAT0017137 | |
| Cricetulus griseus | cgr-miR-181c-3p | MIMAT0023802 | |
| Dasypus novemcinctus | dno-miR-181c-3p | MIMAT0047696 | |
| Goat | chi-miR-181c-3p | MIMAT0036004 | |
| Guinea pig | cpo-miR-181c-3p | MIMAT0047047 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-181c-3p | MIMAT0040008 | |
| Rabbit | ocu-miR-181c-3p | MIMAT0048282 | |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Dasypus novemcinctus
Goat
Guinea pig
Pteropus alecto
Rabbit
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-181c* (v17)
Rat: rno-miR-181c* (v18)
Rat: rno-miR-181c* (v18)

Genomic Map