Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 8 - 84179112 - 84179112 [-] on 84179112 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-181c-5p | MIMAT0000674 |
| Mouse | mmu-miR-181c-3p | MIMAT0017068 |
Stem-loop Details
Mouse

Genomic Map