Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 19 - 25290462 - 25290462 [+] on 25290462 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-181c-5p | MIMAT0000857 |
| Rat | rno-miR-181c-3p | MIMAT0017137 |
Stem-loop Details
Rat

Genomic Map