Assay Details
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCAGCAUUGUACAGGGCUAUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-107 | MIMAT0000104 |
| Mouse | mmu-miR-107-3p | MIMAT0000647 |
| Rat | rno-miR-107-3p | MIMAT0000826 |
| Chicken | gga-miR-107-3p | MIMAT0001147 |
| Dasypus novemcinctus | dno-miR-107-3p | MIMAT0047609 |
| Gorilla gorilla | ggo-miR-107 | MIMAT0002460 |
| View More Rows | ||
| Guinea pig | cpo-miR-107-3p | MIMAT0046960 |
| Horse | eca-miR-107b | MIMAT0012886 |
| Lagothrix lagotricha | lla-miR-107 | MIMAT0002462 |
| Macaca nemestrina | mne-miR-107 | MIMAT0002463 |
| Monodelphis domestica | mdo-miR-107 | MIMAT0004100 |
| Ophiophagus hannah | oha-miR-107-3p | MIMAT0036655 |
| Pan paniscus | ppa-miR-107 | MIMAT0002464 |
| Pan troglodytes | ptr-miR-107 | MIMAT0002459 |
| Pig | ssc-miR-107 | MIMAT0002155 |
| Pongo pygmaeus | ppy-miR-107 | MIMAT0002461 |
| Rabbit | ocu-miR-107-3p | MIMAT0048200 |
| Rhesus monkey | mml-miR-107-3p | MIMAT0002458 |
| Taeniopygia guttata | tgu-miR-107 | MIMAT0014556 |
| Tetraodon nigroviridis | tni-miR-107 | MIMAT0003044 |
| Xenopus tropicalis | xtr-miR-107 | MIMAT0003584 |
| Zebrafish | dre-miR-107a-3p | MIMAT0001817 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-107 |
| Stem-loop Accession # | MI0000114 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | Chr. 10 - 89592747 - 89592827 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000104
miRBase ID: hsa-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: Chr. 10 - 89592747 - 89592827 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-107 |
| Stem-loop Accession # | MI0000684 |
| Stem-loop Sequence | UUCUCUGUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC |
| Chromosome Location | Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000647
miRBase ID: mmu-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-107 |
| Stem-loop Accession # | MI0000890 |
| Stem-loop Sequence | UUCUCUCUGCUUUAAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC |
| Chromosome Location | Chr. 1 - 253128250 - 253128336 [-] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000826
miRBase ID: rno-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: Chr. 1 - 253128250 - 253128336 [-] on Build Rnor_6.0 |
| Stem-loop ID | gga-mir-107 |
| Stem-loop Accession # | MI0001215 |
| Stem-loop Sequence | CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001147
miRBase ID: gga-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | dno-mir-107 |
| Stem-loop Accession # | MI0038962 |
| Stem-loop Sequence | AGCUUCUUUACAGUGUUGCCUUGUGGAAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGAGAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047609
miRBase ID: dno-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ggo-mir-107 |
| Stem-loop Accession # | MI0002756 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002460
miRBase ID: ggo-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-107 |
| Stem-loop Accession # | MI0038620 |
| Stem-loop Sequence | AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046960
miRBase ID: cpo-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | eca-mir-107b |
| Stem-loop Accession # | MI0012643 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012886
miRBase ID: eca-miR-107b Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | lla-mir-107 |
| Stem-loop Accession # | MI0002758 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002462
miRBase ID: lla-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | mne-mir-107 |
| Stem-loop Accession # | MI0002759 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002463
miRBase ID: mne-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | mdo-mir-107 |
| Stem-loop Accession # | MI0005286 |
| Stem-loop Sequence | CUUCUUUCUGCUUUCGGCUUCUCUACAGUGUUGCCUUGUGGCGUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004100
miRBase ID: mdo-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | oha-mir-107 |
| Stem-loop Accession # | MI0031316 |
| Stem-loop Sequence | CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036655
miRBase ID: oha-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ppa-mir-107 |
| Stem-loop Accession # | MI0002760 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002464
miRBase ID: ppa-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-107 |
| Stem-loop Accession # | MI0002755 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002459
miRBase ID: ptr-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ssc-mir-107 |
| Stem-loop Accession # | MI0002449 |
| Stem-loop Sequence | UUCUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002155
miRBase ID: ssc-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-107 |
| Stem-loop Accession # | MI0002757 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002461
miRBase ID: ppy-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | ocu-mir-107 |
| Stem-loop Accession # | MI0039274 |
| Stem-loop Sequence | AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048200
miRBase ID: ocu-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | mml-mir-107 |
| Stem-loop Accession # | MI0002754 |
| Stem-loop Sequence | CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002458
miRBase ID: mml-miR-107-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | tgu-mir-107 |
| Stem-loop Accession # | MI0013773 |
| Stem-loop Sequence | UUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014556
miRBase ID: tgu-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | tni-mir-107 |
| Stem-loop Accession # | MI0003380 |
| Stem-loop Sequence | UCUGUCUGCUGUGAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGCGAUCAAGCAGCAUUGUACAGGGCUAUCACCGCACACAGAGCUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003044
miRBase ID: tni-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | xtr-mir-107 |
| Stem-loop Accession # | MI0004823 |
| Stem-loop Sequence | CUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003584
miRBase ID: xtr-miR-107 Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
| Stem-loop ID | dre-mir-107a |
| Stem-loop Accession # | MI0001963 |
| Stem-loop Sequence | UCUGUGUGCUCUGAGCUUCUUUACAGUGUUGUCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUCACAGCACACUGAACAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001817
miRBase ID: dre-miR-107a-3p Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA Chromosome Location: NA |
More Information
Related Products

Genomic Map