Assay Details
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCAGCAUUGUACAGGGCUAUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-107 | MIMAT0000104 |
| Mouse | mmu-miR-107-3p | MIMAT0000647 |
| Rat | rno-miR-107-3p | MIMAT0000826 |
| Chicken | gga-miR-107-3p | MIMAT0001147 |
| Dasypus novemcinctus | dno-miR-107-3p | MIMAT0047609 |
| Gorilla gorilla | ggo-miR-107 | MIMAT0002460 |
| View More Rows | ||
| Guinea pig | cpo-miR-107-3p | MIMAT0046960 |
| Horse | eca-miR-107b | MIMAT0012886 |
| Lagothrix lagotricha | lla-miR-107 | MIMAT0002462 |
| Macaca nemestrina | mne-miR-107 | MIMAT0002463 |
| Monodelphis domestica | mdo-miR-107 | MIMAT0004100 |
| Ophiophagus hannah | oha-miR-107-3p | MIMAT0036655 |
| Pan paniscus | ppa-miR-107 | MIMAT0002464 |
| Pan troglodytes | ptr-miR-107 | MIMAT0002459 |
| Pig | ssc-miR-107 | MIMAT0002155 |
| Pongo pygmaeus | ppy-miR-107 | MIMAT0002461 |
| Rabbit | ocu-miR-107-3p | MIMAT0048200 |
| Rhesus monkey | mml-miR-107-3p | MIMAT0002458 |
| Taeniopygia guttata | tgu-miR-107 | MIMAT0014556 |
| Tetraodon nigroviridis | tni-miR-107 | MIMAT0003044 |
| Xenopus tropicalis | xtr-miR-107 | MIMAT0003584 |
| Zebrafish | dre-miR-107a-3p | MIMAT0001817 |
Stem-loop Details
Human
Mouse
Rat
Chicken
Dasypus novemcinctus
Gorilla gorilla
Guinea pig
Horse
Lagothrix lagotricha
Macaca nemestrina
Monodelphis domestica
Ophiophagus hannah
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Taeniopygia guttata
Tetraodon nigroviridis
Xenopus tropicalis
Zebrafish

Genomic Map