Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCUUCCCUUUGGACUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520a-3p | MIMAT0002834 |
| Pan troglodytes | ptr-miR-520a | MIMAT0008197 |
| Pongo pygmaeus | ppy-miR-520a-3p | MIMAT0015967 |
| Rhesus monkey | mml-miR-520a-3p | MIMAT0006388 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map