Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCUUCCCUUUGGACUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-520a-3p | MIMAT0002834 |
Pan troglodytes | ptr-miR-520a | MIMAT0008197 |
Pongo pygmaeus | ppy-miR-520a-3p | MIMAT0015967 |
Rhesus monkey | mml-miR-520a-3p | MIMAT0006388 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
