Assay Details
Gene Family ID: | MIPF0000495, mir-920 |
Mature miRNA Sequence: | GGGGAGCUGUGGAAGCAGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-920 | MIMAT0004970 |
| Pan troglodytes | ptr-miR-920 | MIMAT0008336 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map