Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 12 - 24212217 - 24212217 [-] on 24212217 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-920 | MIMAT0004970 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 77 |
Chromosome Location: | Chr. 12 - 24212217 - 24212217 [-] on 24212217 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-920 | MIMAT0004970 |