Assay Details
Gene Family ID: | MIPF0000495, mir-920 |
Mature miRNA Sequence: | GGGGAGCUGUGGAAGCAGUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-920 | MIMAT0004970 |
Pan troglodytes | ptr-miR-920 | MIMAT0008336 |
Stem-loop Details
Human
Pan troglodytes
