Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UAAGUGCUUCCAUGUUUGAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302d-3p | MIMAT0000718 |
| Mouse | mmu-miR-302d-3p | MIMAT0003377 |
| Callithrix jacchus | cja-miR-302d-3p | MIMAT0039590 |
| Dasypus novemcinctus | dno-miR-302d-3p | MIMAT0047798 |
| Pan troglodytes | ptr-miR-302d | MIMAT0008089 |
| Pongo pygmaeus | ppy-miR-302d | MIMAT0015819 |
| View More Rows | ||
| Rabbit | ocu-miR-302d-3p | MIMAT0036323 |
| Rhesus monkey | mml-miR-302d | MIMAT0006262 |
Stem-loop Details
Human
Mouse
Callithrix jacchus
Dasypus novemcinctus
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-302d (v17)
Mouse: mmu-miR-302d (v17)
Mouse: mmu-miR-302d (v17)

Genomic Map