Assay Details
| Gene Family ID: | MIPF0000609, mir-1205 | 
| Mature miRNA Sequence: | UCUGCAGGGUUUGCUUUGAG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-1205 | MIMAT0005869 | 
| Pan troglodytes | ptr-miR-1205 | MIMAT0007959 | 
| Pongo pygmaeus | ppy-miR-1205 | MIMAT0016154 | 
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :