Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 8 - 127960329 - 127960329 [+] on 127960329 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1205 | MIMAT0005869 |
Stem-loop Details
Human
