Assay Details
Gene Family ID: | MIPF0000609, mir-1205 |
Mature miRNA Sequence: | UCUGCAGGGUUUGCUUUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1205 | MIMAT0005869 |
| Pan troglodytes | ptr-miR-1205 | MIMAT0007959 |
| Pongo pygmaeus | ppy-miR-1205 | MIMAT0016154 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map