Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 5 - 168768446 - 168768446 [-] on 168768446 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-218-2-3p | MIMAT0004566 |
| Human | hsa-miR-218-5p | MIMAT0000275 |
Stem-loop Details
Human

Genomic Map