Assay Details
Gene Family ID: | MIPF0000026, mir-218 |
Mature miRNA Sequence: | CAUGGUUCUGUCAAGCACCGCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-218-2-3p | MIMAT0004566 |
| Mouse | mmu-miR-218-2-3p | MIMAT0005444 |
| Rat | rno-miR-218a-2-3p | MIMAT0004740 |
| Dasypus novemcinctus | dno-miR-218-2-3p | MIMAT0047770 |
| Guinea pig | cpo-miR-218-2-3p | MIMAT0047127 |
| Pteropus alecto | pal-miR-218-2-3p | MIMAT0040044 |
| View More Rows | ||
| Rabbit | ocu-miR-218-2-3p | MIMAT0048349 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rabbit

Genomic Map