Assay Details
Gene Family ID: | MIPF0000026, mir-218 |
Mature miRNA Sequence: | CAUGGUUCUGUCAAGCACCGCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-218-2-3p | MIMAT0004566 | |
| Mouse | mmu-miR-218-2-3p | MIMAT0005444 | |
| Rat | rno-miR-218a-2-3p | MIMAT0004740 | |
| Dasypus novemcinctus | dno-miR-218-2-3p | MIMAT0047770 | |
| Guinea pig | cpo-miR-218-2-3p | MIMAT0047127 | |
| Pteropus alecto | pal-miR-218-2-3p | MIMAT0040044 | |
| View More Rows | |||
| Rabbit | ocu-miR-218-2-3p | MIMAT0048349 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rabbit
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Rodent MicroRNA B Cards v3.0TaqMan® Human MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-218-2* (v17)
Mouse: mmu-miR-218-2* (v17)
Rat: rno-miR-218-2* (v16)
Rat: rno-miR-218a-2* (v18)
Rat: rno-miR-218* (v15)
Mouse: mmu-miR-218-2* (v17)
Rat: rno-miR-218-2* (v16)
Rat: rno-miR-218a-2* (v18)
Rat: rno-miR-218* (v15)

Genomic Map