Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 1 - 40754055 - 40754055 [+] on 40754055 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30e-5p | MIMAT0000692 |
| Human | hsa-miR-30e-3p | MIMAT0000693 |
Stem-loop Details
Human

Genomic Map