Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCGGAUGUUUACAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30e-3p | MIMAT0000693 |
| Mouse | mmu-miR-30e-3p | MIMAT0000249 |
| Rat | rno-miR-30e-3p | MIMAT0004720 |
| Alligator mississippiensis | ami-miR-30e-3p | MIMAT0038087 |
| Anolis carolinensis | aca-miR-30e-3p | MIMAT0021929 |
| Chrysemys picta | cpi-miR-30e-3p | MIMAT0037684 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-30e-3p | MIMAT0023901 |
| Dasypus novemcinctus | dno-miR-30e-3p | MIMAT0047566 |
| Daubentonia madagascariensis | dma-miR-30e | MIMAT0049282 |
| Dog | cfa-miR-30e | MIMAT0006627 |
| Guinea pig | cpo-miR-30e-3p | MIMAT0046913 |
| Ophiophagus hannah | oha-miR-30e-3p | MIMAT0036887 |
| Ornithorhynchus anatinus | oan-miR-30e-3p | MIMAT0007204 |
| Pig | ssc-miR-30e-3p | MIMAT0013873 |
| Pteropus alecto | pal-miR-30e-3p | MIMAT0039833 |
| Python bivittatus | pbv-miR-30e-3p | MIMAT0038879 |
| Rabbit | ocu-miR-30e-3p | MIMAT0048160 |
| Taeniopygia guttata | tgu-miR-30a-3p | MIMAT0014535 |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Anolis carolinensis
Chrysemys picta
Cricetulus griseus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Guinea pig
Ophiophagus hannah
Ornithorhynchus anatinus
Pig
Pteropus alecto
Python bivittatus
Rabbit
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Human: hsa-miR-30e* (v17)
Mouse: mmu-miR-30e* (v17)
Rat: rno-miR-30e* (v18)
Aeromonas caviae Ae398: aca-miR-30e* (v18)
Ochrobactrum anthropi: oan-miR-30e* (v18)
Mouse: mmu-miR-30e* (v17)
Rat: rno-miR-30e* (v18)
Aeromonas caviae Ae398: aca-miR-30e* (v18)
Ochrobactrum anthropi: oan-miR-30e* (v18)

Genomic Map