Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUUGACUGGAAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30e-5p | MIMAT0000692 |
| Mouse | mmu-miR-30e-5p | MIMAT0000248 |
| Rat | rno-miR-30e-5p | MIMAT0000805 |
| Horse | eca-miR-30e | MIMAT0012916 |
| Pan troglodytes | ptr-miR-30e | MIMAT0008092 |
| Pongo pygmaeus | ppy-miR-30e | MIMAT0015739 |
| View More Rows | ||
| Rhesus monkey | mml-miR-30e-5p | MIMAT0006172 |
| Xenopus tropicalis | xtr-miR-30e | MIMAT0003678 |
| Zebrafish | dre-miR-30e-5p | MIMAT0001807 |
Stem-loop Details
Human
Mouse
Rat
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Xenopus tropicalis
Zebrafish
More Information
Related Products
miRBase Alias:
Human: hsa-miR-30e (v17)
Mouse: mmu-miR-30e (v17)
Rat: rno-miR-30e (v18)
Danio rerio: dre-miR-30e (v18)
Macaca mulatta: mml-miR-30e (v19)
Mouse: mmu-miR-30e (v17)
Rat: rno-miR-30e (v18)
Danio rerio: dre-miR-30e (v18)
Macaca mulatta: mml-miR-30e (v19)

Genomic Map