Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 14 - 101060376 - 101060376 [-] on 101060376 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-496 | MIMAT0002818 |
Stem-loop Details
Human
