Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UGAGUAUUACAUGGCCAAUCUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-496 | MIMAT0002818 | |
Mouse | mmu-miR-496a-3p | MIMAT0003738 | |
Bovine | bta-miR-496 | MIMAT0009336 | |
Dasypus novemcinctus | dno-miR-496-3p | MIMAT0047933 | |
Dog | cfa-miR-496 | MIMAT0009906 | |
Guinea pig | cpo-miR-496-3p | MIMAT0047294 | |
View More Rows | |||
Horse | eca-miR-496 | MIMAT0013166 | |
Pan troglodytes | ptr-miR-496 | MIMAT0008170 | |
Pongo pygmaeus | ppy-miR-496 | MIMAT0015917 | |
Rabbit | ocu-miR-496-3p | MIMAT0048486 | |
Rhesus monkey | mml-miR-496 | MIMAT0006358 |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Mouse: mmu-miR-496-3p (v18)
Mouse: mmu-miR-496 (v17)
Rat: rno-miR-496 (v15)
Mouse: mmu-miR-496 (v17)
Rat: rno-miR-496 (v15)
