Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UGAGUAUUACAUGGCCAAUCUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-496 | MIMAT0002818 |
Mouse | mmu-miR-496a-3p | MIMAT0003738 |
Bovine | bta-miR-496 | MIMAT0009336 |
Dasypus novemcinctus | dno-miR-496-3p | MIMAT0047933 |
Dog | cfa-miR-496 | MIMAT0009906 |
Guinea pig | cpo-miR-496-3p | MIMAT0047294 |
View More Rows | ||
Horse | eca-miR-496 | MIMAT0013166 |
Pan troglodytes | ptr-miR-496 | MIMAT0008170 |
Pongo pygmaeus | ppy-miR-496 | MIMAT0015917 |
Rabbit | ocu-miR-496-3p | MIMAT0048486 |
Rhesus monkey | mml-miR-496 | MIMAT0006358 |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-496-3p (v18)
Mouse: mmu-miR-496 (v17)
Rat: rno-miR-496 (v15)
Mouse: mmu-miR-496 (v17)
Rat: rno-miR-496 (v15)
