Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 14 - 101043902 - 101043902 [-] on 101043902 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1185-5p | MIMAT0005798 |
| Human | hsa-miR-1185-2-3p | MIMAT0022713 |
Stem-loop Details
Human

Genomic Map