Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAGGAUACCCUUUGUAUGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-1185-5p | MIMAT0005798 | |
| Bovine | bta-miR-1185 | MIMAT0009973 | |
| Horse | eca-miR-1185 | MIMAT0013123 | |
| Pan troglodytes | ptr-miR-1185 | MIMAT0007953 | |
| Pongo pygmaeus | ppy-miR-1185 | MIMAT0016148 | |
| Pteropus alecto | pal-miR-1185-5p | MIMAT0039973 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-1185-5p | MIMAT0024314 | |
Stem-loop Details
Human
Bovine
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

Genomic Map