Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAGGAUACCCUUUGUAUGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1185-5p | MIMAT0005798 |
Bovine | bta-miR-1185 | MIMAT0009973 |
Horse | eca-miR-1185 | MIMAT0013123 |
Pan troglodytes | ptr-miR-1185 | MIMAT0007953 |
Pongo pygmaeus | ppy-miR-1185 | MIMAT0016148 |
Pteropus alecto | pal-miR-1185-5p | MIMAT0039973 |
View More Rows | ||
Rhesus monkey | mml-miR-1185-5p | MIMAT0024314 |
Stem-loop Details
Human
Bovine
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
