Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | ACUAUAUAUCAAGCAUAUUCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-190a-3p | MIMAT0016998 |
Rat | rno-miR-190a-3p | MIMAT0017145 |
Stem-loop Details
Mouse
Rat

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-190-3p (v18)
Mouse: mmu-miR-190* (v17)
Rat: rno-miR-190* (v18)
Mouse: mmu-miR-190* (v17)
Rat: rno-miR-190* (v18)
