Assay Details
Amplicon Length: | 97 |
Chromosome Location: | Chr. 8 - 80927993 - 80927993 [+] on 80927993 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-190a-5p | MIMAT0000865 |
| Rat | rno-miR-190a-3p | MIMAT0017145 |
Stem-loop Details
Rat

Genomic Map