Assay Details
Amplicon Length: | 58 |
Chromosome Location: | Chr. 9 - 67236954 - 67236954 [-] on 67236954 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-190a-5p | MIMAT0000220 |
Mouse | mmu-miR-190a-3p | MIMAT0016998 |
Stem-loop Details
Mouse
