Assay Details
Gene Family ID: | MIPF0000018, mir-154 | 
Mature miRNA Sequence: | AUAUACAGGGGGAGACUCUUAU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-1185-1-3p | MIMAT0022838 | 
| Dog | cfa-miR-1185 | MIMAT0034383 | 
| Guinea pig | cpo-miR-1185-3p | MIMAT0047400 | 
| Pteropus alecto | pal-miR-1185-3p | MIMAT0039974 | 
Stem-loop Details
Human
Dog
Guinea pig
Pteropus alecto

Genomic Map