Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AUAUACAGGGGGAGACUCUUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1185-1-3p | MIMAT0022838 |
| Dog | cfa-miR-1185 | MIMAT0034383 |
| Guinea pig | cpo-miR-1185-3p | MIMAT0047400 |
| Pteropus alecto | pal-miR-1185-3p | MIMAT0039974 |
Stem-loop Details
Human
Dog
Guinea pig
Pteropus alecto

Genomic Map