Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 14 - 101042677 - 101042677 [-] on 101042677 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1185-1-3p | MIMAT0022838 |
Human | hsa-miR-1185-5p | MIMAT0005798 |
Stem-loop Details
Human
