Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | CUAUAUAUCAAACAUAUUCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-190a-3p | MIMAT0026482 |
Chicken | gga-miR-190a-3p | MIMAT0026525 |
Goat | chi-miR-190a-3p | MIMAT0036020 |
Monodelphis domestica | mdo-miR-190a-3p | MIMAT0026933 |
Rhesus monkey | mml-miR-190a-3p | MIMAT0026572 |
Stem-loop Details
Human
Chicken
Goat
Monodelphis domestica
Rhesus monkey
