Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | CUAUAUAUCAAACAUAUUCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-190a-3p | MIMAT0026482 |
| Chicken | gga-miR-190a-3p | MIMAT0026525 |
| Goat | chi-miR-190a-3p | MIMAT0036020 |
| Monodelphis domestica | mdo-miR-190a-3p | MIMAT0026933 |
| Rhesus monkey | mml-miR-190a-3p | MIMAT0026572 |
Stem-loop Details
Human
Chicken
Goat
Monodelphis domestica
Rhesus monkey

Genomic Map