Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 15 - 62823758 - 62823758 [-] on 62823758 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-190a-3p | MIMAT0026482 |
Human | hsa-miR-190a-5p | MIMAT0000458 |
Stem-loop Details
Human
