Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 4 - 120772300 - 120772300 [-] on 120772300 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-30e-5p | MIMAT0000248 |
| Mouse | mmu-miR-30e-3p | MIMAT0000249 |
| Mouse | mmu-miR-30f | MIMAT0025179 |
Stem-loop Details
Mouse

Genomic Map