Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 11 - 35616617 - 35616617 [+] on 35616617 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-218-5p | MIMAT0000663 |
| Mouse | mmu-miR-218-2-3p | MIMAT0005444 |
Stem-loop Details
Mouse

Genomic Map