Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 5 - 139702617 - 139702617 [-] on 139702617 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-30e-3p | MIMAT0004720 |
| Rat | rno-miR-30e-5p | MIMAT0000805 |
Stem-loop Details
Rat

Genomic Map