Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 10 - 20429007 - 20429007 [+] on 20429007 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-218a-2-3p | MIMAT0004740 |
| Rat | rno-miR-218a-5p | MIMAT0000888 |
Stem-loop Details
Rat

Genomic Map